KEY048961879 Key Organics / BIONET

2-(4-bromophenyl)-4-(2-hydroxyethyl)-5-methyl-2,3-dihydro-1H-pyrazol-3-one

  • MDL number MFCD06659582
  • Cas Number : 885949-90-6
  • Molecular Weight : 297.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 171 - 173
Iupac Name 2-(4-bromophenyl)-4-(2-hydroxyethyl)-5-methyl-1,2-dihydro-3H-pyrazol-3-one
Inchi Code 1S/C12H13BrN2O2/c1-8-11(6-7-16)12(17)15(14-8)10-4-2-9(13)3-5-10/h2-5,14,16H,6-7H2,1H3
Inchi Key HBMVCZMODFDTQR-UHFFFAOYSA-N

Description

Melting Point 171 - 173
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88243

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7Y-0201_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart