KEY048961886 Key Organics / BIONET

3-(2,3-dichlorophenyl)-3-(1-oxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

  • MDL number MFCD06659590
  • Cas Number : 885949-95-1
  • Molecular Weight : 350.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209 - 212
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88269

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7Y-0810_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart