KEY048961887 Key Organics / BIONET

5-amino-1-(2,3-dimethylphenyl)-1H-pyrazole-4-carbonitrile

  • MDL number MFCD06659592
  • Cas Number : 792953-00-5
  • Molecular Weight : 212.256

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140 - 142
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88286

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7Y-0951_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart