KEY048961890 Key Organics / BIONET

3-(1,1-dioxo-1lambda6-thiomorpholin-4-yl)benzoic acid

  • MDL number MFCD06659598
  • Cas Number : 763073-96-7
  • Molecular Weight : 255.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 215 - 217
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91352

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8Y-0805_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart