KEY048961892 Key Organics / BIONET

4-methyl-6-(thiophen-3-yl)pyrimidin-2-amine

  • MDL number MFCD06659600
  • Cas Number : 885950-01-6
  • Molecular Weight : 191.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 200 - 203
Iupac Name 4-methyl-6-(3-thienyl)-2-pyrimidinamine
Inchi Code 1S/C9H9N3S/c1-6-4-8(12-9(10)11-6)7-2-3-13-5-7/h2-5H,1H3,(H2,10,11,12)
Inchi Key NHDPFPXZSUELJR-UHFFFAOYSA-N

Description

Melting Point 200 - 203
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91356

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/8Y-0823_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart