KEY048961893 Key Organics / BIONET

2-[(2-chloro-4-nitrophenyl)sulfanyl]acetic acid

  • MDL number MFCD06411624
  • Cas Number : 30880-64-9
  • Molecular Weight : 247.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108 - 110
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81109

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5Y-0803_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart