KEY048961894 Key Organics / BIONET

2-(3-chlorophenyl)quinoline-4-carboxylic acid

  • MDL number MFCD02055964
  • Cas Number : 20389-10-0
  • Molecular Weight : 283.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 214
Iupac Name 2-(3-chlorophenyl)-4-quinolinecarboxylic acid
Inchi Code 1S/C16H10ClNO2/c17-11-5-3-4-10(8-11)15-9-13(16(19)20)12-6-1-2-7-14(12)18-15/h1-9H,(H,19,20)
Inchi Key QPDXYIKQOBZSGQ-UHFFFAOYSA-N

Description

Melting Point 214
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73824

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/3Y-0933_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart