KEY048961925 Key Organics / BIONET

3-(4-chlorophenyl)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid

  • MDL number MFCD01863197
  • Cas Number : 194471-87-9
  • Molecular Weight : 421.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 193 - 195
Iupac Name 3-(4-chlorophenyl)-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-beta-alanine
Inchi Code 1S/C24H20ClNO4/c25-16-11-9-15(10-12-16)22(13-23(27)28)26-24(29)30-14-21-19-7-3-1-5-17(19)18-6-2-4-8-20(18)21/h1-12,21-22H,13-14H2,(H,26,29)(H,27,28)
Inchi Key VDMPMJZDURHZTB-UHFFFAOYSA-N

Description

Melting Point 193 - 195
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84496

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6Y-0928_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart