KEY048961934 Key Organics / BIONET

methyl 4-(3-nitrophenyl)-2,4-dioxobutanoate

  • MDL number MFCD02725697
  • Cas Number : 345617-18-7
  • Molecular Weight : 251.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 111 - 113
Iupac Name methyl 4-(3-nitrophenyl)-2,4-dioxobutanoate
Inchi Code 1S/C11H9NO6/c1-18-11(15)10(14)6-9(13)7-3-2-4-8(5-7)12(16)17/h2-5H,6H2,1H3
Inchi Key QYSCKGUZJUBRDG-UHFFFAOYSA-N

Description

Melting Point 111 - 113
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88248

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/7Y-0618_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart