KEY048961938 Key Organics / BIONET

5-amino-2-(morpholin-4-yl)benzoic acid

  • MDL number MFCD01935677
  • Cas Number : 65989-45-9
  • Molecular Weight : 222.244

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 300+
Purity 95
Storage temp. RT
Physical Form Off-white to Dark Grey Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88255

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/7Y-0703_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart