KEY048961954 Key Organics / BIONET

2-(4-methoxyphenyl)pyridin-3-amine

  • MDL number MFCD06411622
  • Cas Number : 663918-44-3
  • Molecular Weight : 200.241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78 - 80
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81101

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5Y-0701_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart