KEY048961955 Key Organics / BIONET

4-{[2-nitro-4-(trifluoromethyl)phenyl]amino}butanoic acid

  • MDL number MFCD00586806
  • Cas Number : 65783-35-9
  • Molecular Weight : 292.214

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 95 - 98
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91344

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8Y-0702_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart