KEY061970155 Key Organics / BIONET

4-chloro-5-hydroxy-3-(4-methoxyphenoxy)-2,5-dihydrofuran-2-one

  • MDL number MFCD06659532
  • Cas Number : 42190-28-3
  • Molecular Weight : 256.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 103 - 106
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3635

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10Y-0706_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart