KEY061970161 Key Organics / BIONET

tert-butyl 4-[(4-tert-butylphenyl)amino]piperidine-1-carboxylate

  • MDL number MFCD06659533
  • Cas Number : 885949-49-5
  • Molecular Weight : 332.488

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115 - 116
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3655

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10Y-0808_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart