KEY061970163 Key Organics / BIONET

methyl 2-(2,5-dimethyl-1H-pyrrol-1-yl)benzoate

  • MDL number MFCD01125269
  • Cas Number : 83935-44-8
  • Molecular Weight : 229.279

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 86 @0.1 mBar
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3663

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10Y-0826_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart