KEY061970165 Key Organics / BIONET

4-(4-nitrophenyl)-4H,5H,6H,7H-thieno[3,2-c]pyridine

  • MDL number MFCD06797378
  • Cas Number : 213462-01-2
  • Molecular Weight : 260.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108 - 110
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3670

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10Y-0844_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart