KEY061970166 Key Organics / BIONET

methyl 4-(3-methylbutoxy)benzoate

  • MDL number MFCD06659534
  • Cas Number : 408340-71-6
  • Molecular Weight : 222.284

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point oil
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3671

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10Y-0923_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart