KEY061970178 Key Organics / BIONET

4-(5-butyl-1,3,4-oxadiazol-2-yl)aniline

  • MDL number MFCD06797387
  • Cas Number : 100933-82-2
  • Molecular Weight : 217.272

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119 - 121
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/63101

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12Y-0825_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart