KEY061970184 Key Organics / BIONET

2,3,4,5,6-pentafluorophenyl ethene-1-sulfonate

  • MDL number MFCD06659554
  • Cas Number : 452905-58-7
  • Molecular Weight : 274.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 74 @.06 mBar
Iupac Name 2,3,4,5,6-pentafluorophenyl ethylenesulfonate
Inchi Code 1S/C8H3F5O3S/c1-2-17(14,15)16-8-6(12)4(10)3(9)5(11)7(8)13/h2H,1H2
Inchi Key NVXWLLDJCHOIBZ-UHFFFAOYSA-N

Description

Melting Point 74 @.06 mBar
Purity 95%
Physical Form Liquid,Low Melting Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/77410

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/4Y-0324_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart