KEY061970186 Key Organics / BIONET

2-(3-oxo-3,4-dihydro-2H-1,4-benzoxazin-6-yl)acetic acid

  • MDL number MFCD06659563
  • Cas Number : 134997-91-4
  • Molecular Weight : 207.185

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 281 - 283
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/81108

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/5Y-0802_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart