KEY061970187 Key Organics / BIONET

methyl 2-methyl-2-(1H-1,2,4-triazol-1-yl)propanoate

  • MDL number MFCD06659569
  • Cas Number : 885949-78-0
  • Molecular Weight : 169.184

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96 @ 1 mBar
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84456

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6Y-0608_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart