KEY061970189 Key Organics / BIONET

3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-[4-(propan-2-yl)phenyl]propanoic acid hydrate

  • MDL number MFCD06659576
  • Cas Number : 1049746-57-7
  • Molecular Weight : 447.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 183 - 185
Iupac Name N-[(9H-fluoren-9-ylmethoxy)carbonyl]-3-(4-isopropylphenyl)-beta-alanine hydrate
Inchi Code 1S/C27H27NO4.H2O/c1-17(2)18-11-13-19(14-12-18)25(15-26(29)30)28-27(31)32-16-24-22-9-5-3-7-20(22)21-8-4-6-10-23(21)24;/h3-14,17,24-25H,15-16H2,1-2H3,(H,28,31)(H,29,30);1H2
Inchi Key OVHFFLREUYJMMD-UHFFFAOYSA-N

Description

Melting Point 183 - 185
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84498

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/6Y-0933_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart