KEY061970192 Key Organics / BIONET

3-[4-(2-hydroxybenzoyl)-1H-pyrazol-1-yl]benzoic acid

  • MDL number MFCD06659579
  • Cas Number : 959575-06-5
  • Molecular Weight : 308.293

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 232
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/88240

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7Y-0027_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart