KEY061970195 Key Organics / BIONET

tert-butyl 4-(4-nitrobenzoyl)piperazine-1-carboxylate

  • MDL number MFCD06659593
  • Cas Number : 509073-62-5
  • Molecular Weight : 335.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 144
Iupac Name tert-butyl 4-(4-nitrobenzoyl)-1-piperazinecarboxylate
Inchi Code 1S/C16H21N3O5/c1-16(2,3)24-15(21)18-10-8-17(9-11-18)14(20)12-4-6-13(7-5-12)19(22)23/h4-7H,8-11H2,1-3H3
Inchi Key XHGPWZQBCGONPX-UHFFFAOYSA-N

Description

Melting Point 144
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91341

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/8Y-0035_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart