KEY061970198 Key Organics / BIONET

1-[(2-chloro-1,3-thiazol-5-yl)methyl]azepane

  • MDL number MFCD06659601
  • Cas Number : 885950-03-8
  • Molecular Weight : 230.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110 @ 0.5 mBar
Purity 97
Storage temp. Store under N2 at 4°C
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91358

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8Y-0825_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart