KEY061970202 Key Organics / BIONET

1-{3-chloro-4-[(4-methyl-3-nitrophenyl)methoxy]phenyl}ethan-1-one

  • MDL number MFCD06659605
  • Cas Number : 885950-07-2
  • Molecular Weight : 319.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134 - 135
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91366

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8Y-0929_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart