KEY061970204 Key Organics / BIONET

ethyl 8-fluoro-1H,2H,3H,4H,5H-pyrido[4,3-b]indole-2-carboxylate

  • MDL number MFCD02930345
  • Cas Number : 58038-66-7
  • Molecular Weight : 262.284

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172 - 173
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94310

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9Y-0006_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart