KEY061970205 Key Organics / BIONET

6-(2-amino-1,3-thiazol-4-yl)-3,4-dihydro-2H-1,4-benzoxazin-3-one hydrochloride

  • MDL number MFCD06659606
  • Cas Number : 1049742-69-9
  • Molecular Weight : 283.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 304 - 306
Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94313

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/9Y-0217_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart