KEY061970208 Key Organics / BIONET

6-phenylpyridine-3-carbaldehyde

  • MDL number MFCD03086130
  • Cas Number : 63056-20-2
  • Molecular Weight : 183.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 54 - 56
Iupac Name 6-phenylnicotinaldehyde
Inchi Code 1S/C12H9NO/c14-9-10-6-7-12(13-8-10)11-4-2-1-3-5-11/h1-9H
Inchi Key HWDMDOSKSDLXEH-UHFFFAOYSA-N

Description

Melting Point 54 - 56
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94317

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/9Y-0703_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart