KEY061970209 Key Organics / BIONET

2-(thiophen-3-yl)pyridine-3-carbaldehyde

  • MDL number MFCD06659610
  • Cas Number : 885950-13-0
  • Molecular Weight : 189.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60 - 62
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94319

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9Y-0708_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart