KEY061970214 Key Organics / BIONET

2-(chloromethyl)-[1,3]oxazolo[4,5-b]pyridine

  • MDL number MFCD06659615
  • Cas Number : 110704-34-2
  • Molecular Weight : 168.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 113 - 116
Iupac Name 2-(chloromethyl)[1,3]oxazolo[4,5-b]pyridine
Inchi Code 1S/C7H5ClN2O/c8-4-6-10-7-5(11-6)2-1-3-9-7/h1-3H,4H2
Inchi Key MUBURDYXGKXMFA-UHFFFAOYSA-N

Description

Melting Point 113 - 116
Purity 95%
Storage temp. Store under N2, Hygroscopic
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94325

Safety Information

signalword Warning
pcodes P232,P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/9Y-0716_SDS.pdf
hcodes H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart