KEY061970216 Key Organics / BIONET

3-bromoimidazo[1,2-a]pyridine-6-carbonitrile

  • MDL number MFCD06659617
  • Cas Number : 885950-21-0
  • Molecular Weight : 222.045

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 212 - 213
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94331

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9Y-0808_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart