KEY061970217 Key Organics / BIONET

1-(3,4-dibromothiophen-2-yl)ethan-1-one

  • MDL number MFCD06659618
  • Cas Number : 57681-57-9
  • Molecular Weight : 283.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 78 - 80
Purity 95
Storage temp. Store under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94332

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P422
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9Y-0810_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart