KEY061970218 Key Organics / BIONET

N-{3-[3-(morpholin-4-yl)prop-1-yn-1-yl]phenyl}acetamide

  • MDL number MFCD06795474
  • Cas Number : 885950-22-1
  • Molecular Weight : 258.321

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110 - 112
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94336

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/9Y-0820_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart