KEY061970221 Key Organics / BIONET

1-(4,5-dibromothiophen-2-yl)ethan-1-one

  • MDL number MFCD06659622
  • Molecular Weight : 283.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 83 - 85
Iupac Name 1-(4,5-dibromo-2-thienyl)ethanone
Inchi Code 1S/C6H4Br2OS/c1-3(9)5-2-4(7)6(8)10-5/h2H,1H3
Inchi Key OQALQEZJNNWAFM-UHFFFAOYSA-N

Description

Melting Point 83 - 85
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/94341

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/9Y-0839_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart