KEY061970256 Key Organics / BIONET

2-(morpholin-4-yl)pyridine-3-carbaldehyde

  • MDL number MFCD03086195
  • Cas Number : 465514-09-4
  • Molecular Weight : 192.218

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51 - 53
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66498

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1Y-0719_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart