KEY064745008 Key Organics / BIONET

(2E)-2-methanesulfonyl-3-(5-methoxy-1H-indol-2-yl)prop-2-enenitrile

  • MDL number MFCD07021408
  • Cas Number : 900014-99-5
  • Molecular Weight : 276.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2E)-3-(5-methoxy-1H-indol-2-yl)-2-(methylsulfonyl)-2-propenenitrile
Inchi Code 1S/C13H12N2O3S/c1-18-11-3-4-13-9(6-11)5-10(15-13)7-12(8-14)19(2,16)17/h3-7,15H,1-2H3/b12-7+
Inchi Key HIJQBHFXHYJFFM-KPKJPENVSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/73853

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/3Y-5079_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart