KEY064745149 Key Organics / BIONET

ethyl 1-[4-(4-chlorophenyl)-1,3-thiazol-2-yl]-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD00665229
  • Cas Number : 159885-63-9
  • Molecular Weight : 401.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98 - 100
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/85919

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/7K-518S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart