KEY064745166 Key Organics / BIONET

ethyl 6-chloro-2-(trifluoromethyl)imidazo[1,2-a]pyridine-3-carboxylate

  • MDL number MFCD06637317
  • Cas Number : 420130-58-1
  • Molecular Weight : 292.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 100 - 102
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/91273

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/8X-0839_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart