KEY087886551 Key Organics / BIONET

1-benzyl-2-(prop-2-en-1-ylsulfanyl)-1H-imidazole-5-carbaldehyde

  • MDL number MFCD08056602
  • Cas Number : 885950-31-2
  • Molecular Weight : 258.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point OIL
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2097

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10M-344S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart