KEY087886552 Key Organics / BIONET

2,2,2-trichloro-1-(4-cyclohexanecarbonyl-1H-pyrrol-2-yl)ethan-1-one

  • MDL number MFCD07698575
  • Cas Number : 885950-32-3
  • Molecular Weight : 322.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 189 - 191
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/2850

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10R-0331_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart