KEY087886554 Key Organics / BIONET

4-fluoro-4'-methoxy-[1,1'-biphenyl]-3-carbonitrile

  • MDL number MFCD08056603
  • Cas Number : 885950-44-7
  • Molecular Weight : 227.238

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 94 - 96
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3642

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/10Y-0721_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart