KEY087886556 Key Organics / BIONET

3-acetyl-6-tert-butyl-1,2-dihydropyridin-2-one

  • MDL number MFCD08056604
  • Cas Number : 885950-54-9
  • Molecular Weight : 193.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 164 - 166
Iupac Name 3-acetyl-6-tert-butyl-2(1H)-pyridinone
Inchi Code 1S/C11H15NO2/c1-7(13)8-5-6-9(11(2,3)4)12-10(8)14/h5-6H,1-4H3,(H,12,14)
Inchi Key JHUIYLMJMURDCO-UHFFFAOYSA-N

Description

Melting Point 164 - 166
Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3743

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/11A-063_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart