KEY087886558 Key Organics / BIONET

3-nitro-4-({[3-(trifluoromethyl)phenyl]methyl}sulfanyl)benzaldehyde

  • MDL number MFCD07698578
  • Cas Number : 885950-56-1
  • Molecular Weight : 341.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142 - 144
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/3976

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11D-134S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart