KEY087886561 Key Organics / BIONET

2-(4-chlorophenyl)-6-oxo-1-(pentan-3-yl)piperidine-3-carboxylic acid

  • MDL number MFCD08056606
  • Cas Number : 886360-97-0
  • Molecular Weight : 323.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213 - 215
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/7353

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/11Y-0229_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart