KEY087886563 Key Organics / BIONET

methyl 4-{4-[bis(acetyloxy)methyl]-2-nitrophenoxy}benzoate

  • MDL number MFCD08056607
  • Cas Number : 886361-08-6
  • Molecular Weight : 403.343

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 67 - 69
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/9644

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/12R-0296_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart