KEY087886575 Key Organics / BIONET

6-{[(4-chlorophenyl)sulfanyl]methyl}-2-methylpyrimidin-4-ol

  • MDL number MFCD08056612
  • Cas Number : 886361-45-1
  • Molecular Weight : 266.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 168 - 170
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/64705

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1L-712S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart