KEY087886577 Key Organics / BIONET

methyl 4'-nitro-3'-(trifluoromethyl)-[1,1'-biphenyl]-4-carboxylate

  • MDL number MFCD07698580
  • Cas Number : 886361-47-3
  • Molecular Weight : 325.243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143 - 145
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/66500

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/1Y-0728_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart