KEY087886582 Key Organics / BIONET

4-chloro-2-fluoro-5-(pyrimidin-2-yloxy)aniline

  • MDL number MFCD08056616
  • Cas Number : 213675-94-6
  • Molecular Weight : 239.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134 - 136
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/68547

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2N-903_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart