KEY087886586 Key Organics / BIONET

1-(3,4-dichlorophenyl)-2-hydroxyethan-1-one

  • MDL number MFCD07698585
  • Cas Number : 113337-38-5
  • Molecular Weight : 205.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116 - 118
Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/70056

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/2Z-0712_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart